Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfoxide, 99.8+%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals

Catalog No. AC327182500
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327182500 250 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC327182500 Supplier Thermo Scientific Chemicals Supplier No. 327182500
 Restricted Item
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Density 1.1000g/mL
Quantity 250 mL
Flash Point 87°C
Packaging Glass Bottle
Physical Form Liquid at 20°C
Refractive Index 1.4770 to 1.4800
Residue after Evaporation 0.001% max.
Specific Gravity 1.1
Assay Percent Range 99.8+%
Linear Formula (CH3)2SO
Viscosity 1.987 mPa.s (25°C)
Water 0.1% max. (Coulometric)
Beilstein 01, IV, 1277
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Merck Index 15, 3285
Boiling Point 189.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
Formula Weight 78.13
Percent Purity ≥99.8%
Grade Molecular Biology
Chemical Name or Material Methyl sulfoxide
Show More Show Less
Hazard Category HYGROSCOPIC

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.