Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfoxide-d6, for NMR, 100.0 atom % D, min. 99.96 atom % D

Catalog No. AC169020250
Change view
Click to view available options
25mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC169020250 25mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC169020250 Supplier Thermo Scientific Chemicals Supplier No. 169020250
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2206-27-1 | C2H6OS | 84.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Quantity 25 mL
Flash Point 88°C
Viscosity 2.4 mPa.s (20°C)
Specific Gravity 1.19
Water 0.02% max. (Coulometric)
Deuterium (D) 99.95% min.
Density 1.1900g/mL
CAS 2206-27-1
Melting Point 20.2°C
Boiling Point 190.0°C
Molecular Formula C2H6OS
Linear Formula (CD3)2SO
MDL Number MFCD00002090
Beilstein 01, IV, 1279
Fieser 13,124; 16,177
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
Molecular Weight (g/mol) 84.17
PubChem CID 75151
Formula Weight 84.18
Grade NMR
Chemical Name or Material Methyl sulfoxide-d6
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 218-617-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.