Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfoxide-d6,

Catalog No. AC464071000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464071000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC464071000 Supplier Thermo Scientific Chemicals Supplier No. 464071000
Add to Cart

C2D6OS | 84.18 g/

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C2D6OS
Molecular Weight (g/mol) 84.18

Specifications

Flash Point 88°C
Viscosity 2.4 mPa.s (20°C)
Specific Gravity 1.1900
Density 1.1900 g/mL
Quantity 100 mL
Melting Point 20.2°C
Boiling Point 190.0°C
CAS 2206-27-1
Solubility Information Solubility aq. soln.: soluble.
Grade NMR
Chemical Name or Material Methyl sulfoxide-d6
Show More Show Less
EINECSNumber 218-617-0

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.