Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl tert-Butyl Ether MTBE ACS, Greenfield

Catalog No. 502945971
Encompass
Change view
Click to view available options
Metail Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-294-5971 Metail Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-294-5971 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P357000ACSPL05M
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1634-04-4
Molecular Weight (g/mol) 88.15

Specifications

Quantity 5 Gallon
Packaging Metal Pail
Boiling Point 131 °F (55 °C)
Percent Purity 100%
Grade Reagent
Chemical Name or Material Methyl tert-Butyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.