Learn More
Medchemexpress LLC Methyl tetracosanoate | 2442-49-1 | MFCD00009351 | 250 MG
SDP
Catalog No. 5000422074
Encompass_Preferred
Methyl tetracosanoate, also known as Methyl lignocerate, is a fatty acid methyl ester primarily recognized for its anti-diabetic activity. This compound has demonstrated the ability to increase glucose uptake in differentiated 3T3-L1 adipocytes and elevates GLUT4 mRNA expression.
  • Increases glucose uptake on differentiated 3T3-L1 adipocytes
  • Elevates GLUT4 mRNA expression
  • Purity of 98.0%
  • Solid appearance
  • White to off-white color
  • Stable for 3 years at room temperature (solid form)
  • Solutions stable for 2 years at -80°C or 1 year at -20°C
Catalog No. 50-004-22074 Supplier Medchemexpress LLC Supplier No. 250MGHYN8438
Methyl tetracosanoate, also known as Methyl lignocerate, is a fatty acid methyl ester primarily recognized for its anti-diabetic activity. This compound has demonstrated the ability to increase glucose uptake in differentiated 3T3-L1 adipocytes and elevates GLUT4 mRNA expression.
  • Increases glucose uptake on differentiated 3T3-L1 adipocytes
  • Elevates GLUT4 mRNA expression
  • Purity of 98.0%
  • Solid appearance
  • White to off-white color
  • Stable for 3 years at room temperature (solid form)
  • Solutions stable for 2 years at -80°C or 1 year at -20°C