Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Tetrahydrofuran-2-carboxylate 98.0+%, TCI America™
SDP

Catalog No. M175025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M175025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M175025G Supplier TCI America Supplier No. M175025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37443-42-8
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00154541
InChI Key IXHZGHPQQTXOKV-UHFFFAOYSA-N
Synonym Tetrahydrofuran-2-carboxylic Acid Methyl Ester, 2-Tetrahydrofuroic Acid Methyl Ester
PubChem CID 169922
IUPAC Name methyl oxolane-2-carboxylate
SMILES COC(=O)C1CCCO1

Specifications

Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 180°C
Formula Weight 130.14
Percent Purity ≥98.0% (GC)
Chemical Name or Material Methyl Tetrahydrofuran-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.