Learn More
AdipoGen Methyl thiazolyl phenol
SDP
Catalog No. 502560587
Encompass_Preferred
Chemical. CAS 30686-73-8. Formula C10H9NOS. MW 191.25. Synthetic. Potent peroxidase chemiluminescence enhancer for HRP.
Catalog No. 50-256-0587 Supplier AdipoGen Supplier No. CDXM0006M050
Chemical. CAS 30686-73-8. Formula C10H9NOS. MW 191.25. Synthetic. Potent peroxidase chemiluminescence enhancer for HRP.