Learn More
AdipoGen Methyl thiazolyl phenol
SDP
Supplier:  AdipoGen CDXM0006M500
Encompass_Preferred
Chemical. CAS 30686-73-8. Formula C10H9NOS. MW 191.25. Synthetic. Potent peroxidase chemiluminescence enhancer for HRP.
Catalog No. 50-256-0586