Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl Thiourea Ethyl Sulfide Ethyl Silica

Catalog No. 74533250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-533-250G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 74-533-250G Supplier Sigma Aldrich Supplier No. 74533250G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silica Supported Metal Scavenger:Ru(III), Rh(I), Rh(III), Pd(0), Pd(II), Os, Ir(I), Ir(III), Pt(II)Sold in collaboration with PhosphonicS Ltd. for research purposes only.

Specifications

Molecular Formula C18H24NP
MDL Number MFCD17013995
Quantity 500 mg
Molecular Weight (g/mol) 285.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.