Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl trans-2-Octenoate 95.0+%, TCI America™
SDP

Catalog No. M20145ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20145ML 5 mL
M201425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20145ML Supplier TCI America Supplier No. M20145ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7367-81-9
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00036620
InChI Key CJBQSBZJDJHMLF-BQYQJAHWSA-N
Synonym methyl trans-2-octenoate, methyl oct-2-enoate, methyl 2-octenoate, 2-octenoic acid, methyl ester, methyl e-oct-2-enoate, unii-9en9v40j78, 2-octenoic acid, methyl ester, e, 2-octenoic acid, methyl ester, 2e, 2-octenoic acid methyl ester, fema no. 3712
PubChem CID 5364532
IUPAC Name methyl (E)-oct-2-enoate
SMILES CCCCCC=CC(=O)OC

Specifications

Color Colorless
Boiling Point 91°C
Quantity 5 mL
Formula Weight 156.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl trans-2-Octenoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.