Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl trans-4-bromo-2-butenoate

Catalog No. 165055ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-505-5ML 5 mL
16-505-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-505-5ML Supplier Sigma Aldrich Supplier No. 165055ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl trans-4-bromo-2-butenoate (Methyl 4-bromocrotonate) was used in the synthesis of highly substituted benzoxepines or benzopyrans and L-α-amino [4,5-3H]adipic acid.

Specifications

CAS 6000-00-6
Density 1.498 g/mL (at 20°C (literature))
Boiling Point 70°C (0.03 mmHg)
Molecular Formula C5H7BrO2
Linear Formula BrCH2CH=CHCOOCH3
Refractive Index n20/D 1.501
MDL Number MFCD00000246
Quantity 5 mL
Synonym Methyl 4-bromocrotonate
Molecular Weight (g/mol) 179.01
Percent Purity ≥90% (GC)
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.