Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl trans-6-Octadecenoate 97.0+%, TCI America™
SDP

Catalog No. M2310100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2310100MG 100 mg
M2310500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2310100MG Supplier TCI America Supplier No. M2310100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14620-36-1
Molecular Formula C19H36O2
Molecular Weight (g/mol) 296.495
MDL Number MFCD00069607
InChI Key QRTVDKVXAFJVRU-BUHFOSPRSA-N
Synonym trans-6-Octadecenoic Acid Methyl Ester, Methyl Petroselaidate, Petroselaidic Acid Methyl Ester
PubChem CID 5366845
IUPAC Name methyl (E)-octadec-6-enoate
SMILES CCCCCCCCCCCC=CCCCCC(=O)OC

Specifications

Melting Point 19°C
Color Colorless
Boiling Point 165°C
Quantity 100 mg
Formula Weight 296.50
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl trans-6-Octadecenoate
EINECSNumber (2)-0798
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.