Learn More
Medchemexpress LLC Methyl trans-p-coumarate | 19367-38-5 | 50 G
SDP
Catalog No. 5000421121
Encompass_Preferred
(E)-Methyl 4-coumarate (Methyl 4-hydroxycinnamate) is a phenolic compound and a derivative of Cinnamic acid. It is found in plants like Allium cepa and Morinda citrifolia L.. This compound induces apoptosis when combined with Carnosic acid, inhibits GSK3β activity, modulates inflammatory cytokine levels by increasing IL-10 and decreasing IL-4, and shows anticancer effects against acute myeloid leukemia. It can also ameliorate Plasmodium berghei NK65 infection.
  • Phenolic compound and derivative of cinnamic acid.
  • Found in Allium cepa and Morinda citrifolia L..
  • Induces apoptosis when combined with carnosic acid.
  • Inhibits GSK3β activity.
  • Modulates inflammatory cytokine levels (increases IL-10, decreases IL-4).
  • Exhibits anticancer effects against acute myeloid leukemia in combination with carnosic acid.
  • Ameliorates Plasmodium berghei NK65 infection.
Catalog No. 50-004-21121 Supplier Medchemexpress LLC Supplier No. HYN249250G
(E)-Methyl 4-coumarate (Methyl 4-hydroxycinnamate) is a phenolic compound and a derivative of Cinnamic acid. It is found in plants like Allium cepa and Morinda citrifolia L.. This compound induces apoptosis when combined with Carnosic acid, inhibits GSK3β activity, modulates inflammatory cytokine levels by increasing IL-10 and decreasing IL-4, and shows anticancer effects against acute myeloid leukemia. It can also ameliorate Plasmodium berghei NK65 infection.
  • Phenolic compound and derivative of cinnamic acid.
  • Found in Allium cepa and Morinda citrifolia L..
  • Induces apoptosis when combined with carnosic acid.
  • Inhibits GSK3β activity.
  • Modulates inflammatory cytokine levels (increases IL-10, decreases IL-4).
  • Exhibits anticancer effects against acute myeloid leukemia in combination with carnosic acid.
  • Ameliorates Plasmodium berghei NK65 infection.