Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Tricosafluorododecanoate 95.0+%, TCI America™
SDP

Catalog No. M19175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19175G Supplier TCI America Supplier No. M19175G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56554-52-0
Molecular Formula C13H3F23O2
Molecular Weight (g/mol) 628.128
MDL Number MFCD00236616
InChI Key ABNZIVVYHQYSLI-UHFFFAOYSA-N
Synonym methyl perfluorododecanoate, methyl tricosafluorododecanoate, tricosafluorododecanoic acid methyl ester, dodecanoic acid, tricosafluoro-, methyl ester, methyl tricosafluorolaurate, acmc-1b030, perfluorododecanoic acid methyl ester, methyl tricosafluorolaurate; perfluorododecanoic acid methyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid methyl ester
PubChem CID 547885
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoate
SMILES COC(=O)C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 58°C
Color White-Yellow
Boiling Point 125°C
Quantity 5 g
Formula Weight 628.13
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Tricosafluorododecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.