Learn More
Apexbio Technology LLC Methyl tridecanoate 1731-88-0 10g
SDP
Catalog No. 5000096138
Encompass_Preferred
Methyl tridecanoate (CAS 1731-88-0) is a small-molecule inhibitor targeting -amyloid protein aggregation and acetylcholinesterase (AChE) It is designed to moderately inhibit -amyloid aggregation and weakly inhibit AChE activity Methyl tridecanoate exerts its biological activity primarily through inhibition of -amyloid aggregation and AChE enzymatic function [No experimental model/cell line or IC50/EC50 reported ]Based on these pharmacological properties methyl tridecanoate holds research potential in neurodegenerative disease models and lipid metabolism studies
Catalog No. 50-000-96138 Supplier Apexbio Technology LLC Supplier No. A993210000
Methyl tridecanoate (CAS 1731-88-0) is a small-molecule inhibitor targeting -amyloid protein aggregation and acetylcholinesterase (AChE) It is designed to moderately inhibit -amyloid aggregation and weakly inhibit AChE activity Methyl tridecanoate exerts its biological activity primarily through inhibition of -amyloid aggregation and AChE enzymatic function [No experimental model/cell line or IC50/EC50 reported ]Based on these pharmacological properties methyl tridecanoate holds research potential in neurodegenerative disease models and lipid metabolism studies