Learn More
Medchemexpress LLC Methyl tridecanoate | 1731-88-0 | 228.37 | 5 G
SDP
Catalog No. 5000449203
Encompass_Preferred
Methyl tridecanoate moderately inhibits β-amyloid aggregation. It also weakly inhibits acetylcholinesterase (AChE). This product appears as a colorless to light yellow liquid.
  • Moderately inhibits β-amyloid aggregation.
  • Weakly inhibits acetylcholinesterase (AChE).
  • Available in different forms and quantities for research use.
Catalog No. 50-004-49203 Supplier Medchemexpress LLC Supplier No. HYW0042875G
Methyl tridecanoate moderately inhibits β-amyloid aggregation. It also weakly inhibits acetylcholinesterase (AChE). This product appears as a colorless to light yellow liquid.
  • Moderately inhibits β-amyloid aggregation.
  • Weakly inhibits acetylcholinesterase (AChE).
  • Available in different forms and quantities for research use.