Learn More
Indofine Chemical Methyl Trifluoroace, 95%, 5 Gm
SDP
Catalog No. 50910042
Encompass_Preferred
METHYL TRIFLUOROACETOACETATE 5 GM, Fluoro Organics, Methyl-4,4,4-trifluoroacetoacetate, C5H5F3O3, MW: 170.09, 95+%, 83643-84-9, MFCD00041004
Catalog No. 50-910-042 Supplier Indofine Chemical Supplier No. 091671 5 GM
METHYL TRIFLUOROACETOACETATE 5 GM, Fluoro Organics, Methyl-4,4,4-trifluoroacetoacetate, C5H5F3O3, MW: 170.09, 95+%, 83643-84-9, MFCD00041004