Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl trifluoropyruvate, 97%

Catalog No. AAB2158906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2158906 5 g
AAB2158903 1 g
AAB2158914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2158906 Supplier Thermo Scientific Chemicals Supplier No. B2158906
Only null left
Add to cart

CAS: 13089-11-7 | C4H3F3O3 | 156.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13089-11-7
Molecular Formula C4H3F3O3
Molecular Weight (g/mol) 156.06
MDL Number MFCD00114936
InChI Key XGLLQDIWQRQROJ-UHFFFAOYSA-N
Synonym methyl trifluoropyruvate, methyl 3,3,3-trifluoropyruvate, methyltrifluoropyruvate, trifluoropyruvic acid methyl ester, 3,3,3-trifluoropyruvic acid methyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, methyl ester, pubchem2012, rarechem al bf 1300, acmc-1bp0j
PubChem CID 2734931
IUPAC Name methyl 3,3,3-trifluoro-2-oxopropanoate
SMILES COC(=O)C(=O)C(F)(F)F

Specifications

Density 1.52
Boiling Point 84°C to 86°C
Flash Point 26°C (79°F)
Refractive Index 1.332
Quantity 5 g
UN Number UN3272
Beilstein 1863490
Formula Weight 156.06
Percent Purity 97%
Chemical Name or Material Methyl trifluoropyruvate
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.