Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Trifluoropyruvate 98.0+%, TCI America™
SDP

Catalog No. T17851G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17851G 1 g
T17855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17851G Supplier TCI America Supplier No. T17851G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13089-11-7
Molecular Formula C4H3F3O3
Molecular Weight (g/mol) 156.06
MDL Number MFCD00114936
InChI Key XGLLQDIWQRQROJ-UHFFFAOYSA-N
Synonym methyl trifluoropyruvate, methyl 3,3,3-trifluoropyruvate, methyltrifluoropyruvate, trifluoropyruvic acid methyl ester, 3,3,3-trifluoropyruvic acid methyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, methyl ester, pubchem2012, rarechem al bf 1300, acmc-1bp0j
PubChem CID 2734931
IUPAC Name methyl 3,3,3-trifluoro-2-oxopropanoate
SMILES COC(=O)C(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 86°C
Quantity 1 g
UN Number 3272
Formula Weight 156.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Trifluoropyruvate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.