Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl trimethylacetate

Catalog No. M86502500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M86502500ML 500 mL
M86502100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M86502500ML Supplier Sigma Aldrich Supplier No. M86502500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl trimethylacetate

Specifications

CAS 598-98-1
Density 0.873 g/mL (at 25°C (literature))
Boiling Point 101°C (lit.)
Molecular Formula C6H12O2
Refractive Index n20/D 1.390 (literature)
Linear Formula (CH3)3 CCOOCH3
MDL Number MFCD00008843
Quantity 500 mL
Synonym Methyl pivalate; Methyl 2,2-dimethylpropionate; Methyl trimethylacetate
Molecular Weight (g/mol) 116.16
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.