Learn More
Medchemexpress LLC Methyl trioctyl ammonium chloride | 5137-55-3 | MFCD00011862 | 98.0% | 404.16 g·mol⁻¹ | C25H54ClN | 1kg
SDP
Catalog No. 5000269164
Encompass_Preferred
Methyl trioctyl ammonium chloride is a quaternary ammonium salt commonly used as a surfactant stabilizer and phase transfer catalyst in various chemical reactions especially in organic synthesis Methyl trioctyl ammonium chloride has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes
Catalog No. 50-002-69164 Supplier Medchemexpress LLC Supplier No. HYY00801KG
Methyl trioctyl ammonium chloride is a quaternary ammonium salt commonly used as a surfactant stabilizer and phase transfer catalyst in various chemical reactions especially in organic synthesis Methyl trioctyl ammonium chloride has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes