Learn More
Medchemexpress LLC Methyl trioctyl ammonium chloride | 5137-55-3 | MFCD00011862 | 98.0% | 404.16 g/mol | C25H54ClN | 50g
SDP
Catalog No. 5000268349
Encompass_Preferred
Methyl trioctyl ammonium chloride is a quaternary ammonium salt commonly used as a surfactant stabilizer and phase transfer catalyst in various chemical reactions especially in organic synthesis Methyl trioctyl ammonium chloride has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes
Catalog No. 50-002-68349 Supplier Medchemexpress LLC Supplier No. HYY008050G
Methyl trioctyl ammonium chloride is a quaternary ammonium salt commonly used as a surfactant stabilizer and phase transfer catalyst in various chemical reactions especially in organic synthesis Methyl trioctyl ammonium chloride has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes