Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl-Trioctylammonium Bis(Trifluoromethylsulfonyl)Imide

Catalog No. 007975GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
00-797-5GF 5 g
00-797-50GF 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 00-797-5GF Supplier Sigma Aldrich Supplier No. 007975GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl-Trioctylammonium Bis(Trifluoromethylsulfonyl)Imide

Specifications

CAS 375395-33-8
Molecular Formula C27H54F6N2O4S2
Refractive Index n20/D 1.439
Linear Formula C27H54F6N2O4S2
MDL Number MFCD08275364
Quantity 5 g
Synonym Methyltri-n-octylammonium bis(trifluoromethanesulfonyl)imide; Trioctylmethylammonium bis(trifluoromethylsulfonyl)imide
Molecular Weight (g/mol) 648.85 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.