Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl undec-10-enoate | 111-81-9 | 99.0% | 198.30 g/mol | C12H22O2 | 100 G
SDP

Catalog No. 5000210399
Encompass_Preferred

Methyl undec-10-enoate is an unsaturated fatty acid methyl ester used as a research reagent and chemical intermediate in flavor, fragrance, and polymer synthesis applications. It is supplied as a colorless to light yellow liquid with a molecular weight of 198.30 g/mol and a reported purity of 99.03%.

  • Unsaturated fatty acid methyl ester used in organic synthesis
  • High purity suitable for research applications
  • Liquid appearance, colorless to light yellow
  • Density approximately 0.882 g/cm3
  • Storage recommendations: pure form at -20°C for long-term stability

Catalog No. 50-002-10399 Supplier Medchemexpress LLC Supplier No. HYW068214100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl undec-10-enoate is an unsaturated fatty acid methyl ester used as a research reagent and chemical intermediate in flavor, fragrance, and polymer synthesis applications. It is supplied as a colorless to light yellow liquid with a molecular weight of 198.30 g/mol and a reported purity of 99.03%.

  • Unsaturated fatty acid methyl ester used in organic synthesis
  • High purity suitable for research applications
  • Liquid appearance, colorless to light yellow
  • Density approximately 0.882 g/cm3
  • Storage recommendations: pure form at -20°C for long-term stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.