Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl undecanoate | 1731-86-8 | 200.32 | 1 G
SDP

Catalog No. 5000445459
Encompass_Preferred

Methyl undecanoate is a laboratory chemical primarily used as an internal standard in gas-liquid chromatography. Intended for research use only, this product requires handling by qualified scientists in equipped facilities.

  • Purity of 98.66% by GC
  • Colorless to off-white liquid appearance
  • Molecular formula C12H24O2
  • CAS number 1731-86-8
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at ≥ 10 mg/mL

Catalog No. 50-004-45459 Supplier Medchemexpress LLC Supplier No. HYW0042851G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl undecanoate is a laboratory chemical primarily used as an internal standard in gas-liquid chromatography. Intended for research use only, this product requires handling by qualified scientists in equipped facilities.

  • Purity of 98.66% by GC
  • Colorless to off-white liquid appearance
  • Molecular formula C12H24O2
  • CAS number 1731-86-8
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at ≥ 10 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.