Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl undecanoate | 1731-86-8 | 200.32 | 10 G
SDP

Catalog No. 5000449206
Encompass_Preferred

Methyl undecanoate is an internal standard used in gas-liquid chromatography. This product has been tested and complies with given specifications, and is intended for research use only.

  • Internal standard for gas-liquid chromatography
  • Purity of 98.7%
  • Molecular weight of 200.32
  • Colorless to off-white liquid appearance
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-49206 Supplier Medchemexpress LLC Supplier No. HYW00428510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl undecanoate is an internal standard used in gas-liquid chromatography. This product has been tested and complies with given specifications, and is intended for research use only.

  • Internal standard for gas-liquid chromatography
  • Purity of 98.7%
  • Molecular weight of 200.32
  • Colorless to off-white liquid appearance
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.