Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl undecanoate | 1731-86-8 | 200.32 | 100 G
SDP

Catalog No. 5000445876
Encompass_Preferred

Methyl undecanoate is a colorless to off-white liquid primarily used as an internal standard in gas-liquid chromatography. With a purity of 98.66%, this compound is for research use only and not intended for patient use.

  • Internal standard for gas-liquid chromatography
  • High purity of 98.66%
  • Colorless to off-white liquid appearance
  • Molecular formula C12H24O2
  • Soluble in DMSO at ≥ 10 mg/mL
  • Stable for up to 3 years when stored at -20°C (pure form)

Catalog No. 50-004-45876 Supplier Medchemexpress LLC Supplier No. HYW004285100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl undecanoate is a colorless to off-white liquid primarily used as an internal standard in gas-liquid chromatography. With a purity of 98.66%, this compound is for research use only and not intended for patient use.

  • Internal standard for gas-liquid chromatography
  • High purity of 98.66%
  • Colorless to off-white liquid appearance
  • Molecular formula C12H24O2
  • Soluble in DMSO at ≥ 10 mg/mL
  • Stable for up to 3 years when stored at -20°C (pure form)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.