Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl undecanoate | 1731-86-8 | 200.32 | 25 G
SDP

Catalog No. 5000448279
Encompass_Preferred

Methyl undecanoate is an internal standard used in gas-liquid chromatography. It is intended for research use only.

  • Purity of 98.66%
  • Appears as a colorless to off-white liquid
  • Molecular formula is C12H24O2
  • Molecular weight is 200.32
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at ≥ 10 mg/mL

Catalog No. 50-004-48279 Supplier Medchemexpress LLC Supplier No. 25GHYW004285
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl undecanoate is an internal standard used in gas-liquid chromatography. It is intended for research use only.

  • Purity of 98.66%
  • Appears as a colorless to off-white liquid
  • Molecular formula is C12H24O2
  • Molecular weight is 200.32
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at ≥ 10 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.