Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl undecanoate | 1731-86-8 | 200.32 | 50 G
SDP

Catalog No. 5000445389
Encompass_Preferred

Methyl undecanoate is a compound utilized as an internal standard in gas-liquid chromatogram applications. It is intended for research use only and not for human use.

  • Purity: 98.66%
  • Appearance: Liquid
  • Color: Colorless to off-white
  • Molecular formula: C12H24O2
  • Initial source: Derived from plants, specifically Sterculia lychnophera Hance within the Sterculiaceae family
  • Shipping: Room temperature within the continental United States
  • Storage (pure form): -20°C for 3 years; 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month
  • Solubility: ≥ 10 mg/mL (49.92 mM) in DMSO

Catalog No. 50-004-45389 Supplier Medchemexpress LLC Supplier No. HYW00428550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl undecanoate is a compound utilized as an internal standard in gas-liquid chromatogram applications. It is intended for research use only and not for human use.

  • Purity: 98.66%
  • Appearance: Liquid
  • Color: Colorless to off-white
  • Molecular formula: C12H24O2
  • Initial source: Derived from plants, specifically Sterculia lychnophera Hance within the Sterculiaceae family
  • Shipping: Room temperature within the continental United States
  • Storage (pure form): -20°C for 3 years; 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month
  • Solubility: ≥ 10 mg/mL (49.92 mM) in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.