Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl undecanoate | 1731-86-8 | MFCD00008957 | 200.32 | 10 MM * 1 ML
SDP

Catalog No. 5000448738
Encompass_Preferred

Methyl undecanoate is an internal standard used in gas-liquid chromatography. This product is intended for research use only and is supplied as a colorless to off-white liquid solution.

  • High purity: 98.7%
  • Molecular weight: 200.32
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-48738 Supplier Medchemexpress LLC Supplier No. HYW00428510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl undecanoate is an internal standard used in gas-liquid chromatography. This product is intended for research use only and is supplied as a colorless to off-white liquid solution.

  • High purity: 98.7%
  • Molecular weight: 200.32
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.