Learn More
Medchemexpress LLC Methyl vanillate | 3943-74-6 | 99.88% | C9H10O4 | 25 G
SDP
Catalog No. 5000357627
Encompass_Preferred
Methyl vanillate is a Wnt/β-catenin pathway activator and a benzoate ester of vanillic acid. Found in Oryza sativa Linn., it functions as an antioxidant and plant metabolite, with a high purity of 99.88%.
  • Wnt/β-catenin pathway activator
  • Antioxidant and plant metabolite
  • Found in Oryza sativa Linn.
  • High purity of 99.88%
  • Increases β-catenin expression and nuclear translocation in calvarial osteoblasts
  • Elevates ALP activity in calvarial osteoblasts
Catalog No. 50-003-57627 Supplier Medchemexpress LLC Supplier No. 25GHY75342
Methyl vanillate is a Wnt/β-catenin pathway activator and a benzoate ester of vanillic acid. Found in Oryza sativa Linn., it functions as an antioxidant and plant metabolite, with a high purity of 99.88%.
  • Wnt/β-catenin pathway activator
  • Antioxidant and plant metabolite
  • Found in Oryza sativa Linn.
  • High purity of 99.88%
  • Increases β-catenin expression and nuclear translocation in calvarial osteoblasts
  • Elevates ALP activity in calvarial osteoblasts