Learn More
Medchemexpress LLC Methyl vanillate | 3943-74-6 | 99.88% | C9H10O4 | 50 G
SDP
Catalog No. 5000433830
Encompass_Preferred
Methyl vanillate, an ingredient in Oryza sativa Linn., functions as a Wnt/β-catenin pathway activator. It is a benzoate ester, specifically the methyl ester of vanillic acid, and is characterized as an antioxidant and a plant metabolite.
  • Wnt/β-catenin pathway activator
  • Benzoate ester
  • Antioxidant
  • Plant metabolite
  • Off-white to light brown solid
  • Soluble in DMSO and H2O
  • Increases expression and nuclear translocation of β-catenin
  • Elevates ALP activity in calvarial osteoblasts
Catalog No. 50-004-33830 Supplier Medchemexpress LLC Supplier No. HY7534250G
Add to Cart
Add to Cart
Methyl vanillate, an ingredient in Oryza sativa Linn., functions as a Wnt/β-catenin pathway activator. It is a benzoate ester, specifically the methyl ester of vanillic acid, and is characterized as an antioxidant and a plant metabolite.
  • Wnt/β-catenin pathway activator
  • Benzoate ester
  • Antioxidant
  • Plant metabolite
  • Off-white to light brown solid
  • Soluble in DMSO and H2O
  • Increases expression and nuclear translocation of β-catenin
  • Elevates ALP activity in calvarial osteoblasts