Learn More
Medchemexpress LLC Methyl vanillate | 3943-74-6 | MFCD00008438 | 99.9% | C9H10O4 | 5 G
SDP
Catalog No. 5000434031
Encompass_Preferred
Methyl vanillate, a benzoate ester and plant metabolite found in Oryza sativa Linn., acts as a Wnt/β-catenin pathway activator. It also functions as an antioxidant and is intended for research use only. This compound exhibits diverse biological activities, making it valuable for various research applications.
  • Acts as a Wnt/β-catenin pathway activator
  • Functions as an antioxidant and plant metabolite
  • Derived from Oryza sativa Linn.
  • Enhances expression and nuclear translocation of β-catenin
  • Elevates ALP activity in calvarial osteoblasts
  • Demonstrates anticancer activity in some cell lines
  • Exhibits anti-allergic activity
Catalog No. 50-004-34031 Supplier Medchemexpress LLC Supplier No. HY753425G
Methyl vanillate, a benzoate ester and plant metabolite found in Oryza sativa Linn., acts as a Wnt/β-catenin pathway activator. It also functions as an antioxidant and is intended for research use only. This compound exhibits diverse biological activities, making it valuable for various research applications.
  • Acts as a Wnt/β-catenin pathway activator
  • Functions as an antioxidant and plant metabolite
  • Derived from Oryza sativa Linn.
  • Enhances expression and nuclear translocation of β-catenin
  • Elevates ALP activity in calvarial osteoblasts
  • Demonstrates anticancer activity in some cell lines
  • Exhibits anti-allergic activity