Learn More
Selleck Chemical LLC Methyl Vanillate S3630-1g
SDP
Supplier:  Selleck Chemical LLC S36301G
Encompass_Preferred
Methyl Vanillate one of the ingredients in Hovenia dulcis Thunb activates the Wnt/ -catenin pathway and induces osteoblast differentiation in vitro
Catalog No. 50-310-8967