Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl vinyl sulfone, 95%, stabilized

Catalog No. AC306730100
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC306730100 10 g Glass bottle
AC306730010 1 g Glass bottle
AC306730500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC306730100 Supplier Thermo Scientific Chemicals Supplier No. 306730100
Only null left
Add to Cart

CAS: 3680-02-2 | C3H6O2S | 106.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3680-02-2
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.14
MDL Number MFCD00007567
InChI Key WUIJTQZXUURFQU-UHFFFAOYSA-N
SMILES CS(=O)(=O)C=C

Specifications

Color Colorless to Yellow
Density 1.2100g/mL
Boiling Point 115.0°C to 117.0°C (19.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4610 to 1.4630
Linear Formula H2C=CHSO2CH3
Quantity 10 g
Specific Gravity 1.21
Synonym methyl vinyl sulfone, ethene, methylsulfonyl, methyl vinyl sulphone, methylvinylsulfone, methanesulfonylethene, sulfone, methyl vinyl, vinyl methyl sulfone, methanesulfonyl-ethene, methylsulfonyl ethene, methyl vinyl sulfphone
IUPAC Name 1-methylsulfonylethene
Stabilizer 150ppm 4-tert.-butylpenol
PubChem CID 19354
Formula Weight 106.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl vinyl sulfone, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Fatal in contact with skin.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-958-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.