Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Violet 2B, pure, certified

Catalog No. p-3776823
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415070250 25 g Glass bottle
AC415071000 100 g Glass bottle
AC415075000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC415070250 Supplier Thermo Scientific Chemicals Supplier No. 415070250
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 8004-87-3 | C24H28ClN3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8004-87-3
Molecular Formula C24H28ClN3
MDL Number MFCD00012568
Synonym Basic Violet 1, C.I. 42535

Specifications

Melting Point 137.0°C
Absorption Ratio 1.06 to 1.19
Dye Content 75% min.
Infrared Spectrum Authentic
Lambda Max 583 to 587nm
Packaging Glass bottle
Beilstein 13, 755
Color Green
Solubility Information Solubility in water: soluble. Other solubilities: soluble in conc. sulfuric acid and nitric acid, soluble in chloroform, partially soluble in alcohol and glycerol
Quantity 25 g
Physical Form Crystalline Powder With Metal Luster
Formula Weight 393.95
Grade Pure
Chemical Name or Material Methyl Violet 2B, Pure
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.