Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Methyl Violet 2B, Certified, C.I. DcMv-5 25 GR
SDP

Catalog No. 5027166
Encompass_Preferred

(Basic Violet 1; Dahlia B; Gentian Violet; Paris Violet; Pyoktanium Coeruleum)
CAS #8004-87-3 C₂₄H₂₈N₄Cl F.W. 393.95
Solubility: 9.0% Water; 8.25% Alcohol; 9.0% Cellosolve; 8.5% Glycol;0.01% Xylene

Used with Methyl Green to stain Epoxy-embedded sections.

Catalog No. 50-271-66 Supplier Electron Microscopy Sciences Supplier No. 18850
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Basic Violet 1; Dahlia B; Gentian Violet; Paris Violet; Pyoktanium Coeruleum)
CAS #8004-87-3 C₂₄H₂₈N₄Cl F.W. 393.95
Solubility: 9.0% Water; 8.25% Alcohol; 9.0% Cellosolve; 8.5% Glycol;0.01% Xylene

Used with Methyl Green to stain Epoxy-embedded sections.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.