Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Violet Indicator, 0.02% (w/v), Ricca Chemical
SDP

Catalog No. R51000001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R51000001A 1 L Natural Poly Bottle
5100-4 120 mL Natural Poly Bottle
5100-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R51000001A Supplier Ricca Chemical Company Supplier No. 510032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 8004-87-3

Chemical Identifiers

CAS 8004-87-3 , 7732-18-5

Specifications

Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Purple
pH >1.6
Quantity 1 L
Percent Purity 0.02% W/V
Grade Indicator
Chemical Name or Material Methyl Violet Indicator, 0.02% (w/v) Aqueous Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.