Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Violet, Indicator (Basic Violet 1; Methyl Violet 2B), Reagents

Catalog No. p-200021427
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-734 10 g
01-336-733 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-734 Supplier Reagents Holdings Llc Supplier No. C611900010C1
Only null left
Add to Cart
Add to Cart

Methyl Violet, Indicator (Basic Violet 1; Methyl Violet 2B), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Methyl Violet, Indicator (Basic Violet 1, Methyl Violet 2B)
CAS 8004-87-3
Molecular Formula C24H27N3.HCl
Synonym Basic Violet 1, N-(4-(bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)methanaminium chloride)
Molecular Weight (g/mol) 393.95
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0395
Color Blue/Green
Quantity 10 g
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.