Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl-warfarin-d3 | 38063-51-3 | 98.0% | 325.37 g·mol⁻1 | C20H15D3O4 | 10 MG
SDP

Catalog No. 5000288645
Encompass_Preferred

Methyl-warfarin-d3 is a deuterium-labeled methylated analog of warfarin provided as a research-use-only stable isotope reference standard for analytical and method-development applications.

  • Deuterium-labeled methyl analog useful as an internal standard.
  • High purity suitable for analytical use (≈98.0%).
  • Molecular formula C20H15D3O4; molecular weight ~325.37 g·mol⁻1.
  • Intended applications: LC-MS, GC-MS, NMR, and method development.
  • Supplied as a 10 mg solid for laboratory use (research use only).

Catalog No. 50-002-88645 Supplier Medchemexpress LLC Supplier No. HY159002S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl-warfarin-d3 is a deuterium-labeled methylated analog of warfarin provided as a research-use-only stable isotope reference standard for analytical and method-development applications.

  • Deuterium-labeled methyl analog useful as an internal standard.
  • High purity suitable for analytical use (≈98.0%).
  • Molecular formula C20H15D3O4; molecular weight ~325.37 g·mol⁻1.
  • Intended applications: LC-MS, GC-MS, NMR, and method development.
  • Supplied as a 10 mg solid for laboratory use (research use only).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.