Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl yellow | 60-11-7 | MFCD00008308 | 225.29 g/mol | C14H15N3 | 25 G
SDP

Catalog No. 5000302212
Encompass_Preferred

Methyl yellow is an azo dye used as a pH indicator that appears red in acidic solutions and turns yellow above pH 4.0. Supplied as a light-yellow solid for laboratory research, it includes handling guidance to protect from light and store refrigerated; solution stability data are provided for long-term storage at low temperatures.

  • pH indicator: red at low pH, yellow above pH 4.0.
  • Chemical formula C14H15N3; molecular weight 225.29 g/mol.
  • Supplied as a light-yellow solid suitable for laboratory handling.
  • Available in common laboratory package sizes (25 g, 50 g).
  • Storage guidance: store at 4°C and protect from light; in solution store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-02212 Supplier Medchemexpress LLC Supplier No. 25GHYW025785
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl yellow is an azo dye used as a pH indicator that appears red in acidic solutions and turns yellow above pH 4.0. Supplied as a light-yellow solid for laboratory research, it includes handling guidance to protect from light and store refrigerated; solution stability data are provided for long-term storage at low temperatures.

  • pH indicator: red at low pH, yellow above pH 4.0.
  • Chemical formula C14H15N3; molecular weight 225.29 g/mol.
  • Supplied as a light-yellow solid suitable for laboratory handling.
  • Available in common laboratory package sizes (25 g, 50 g).
  • Storage guidance: store at 4°C and protect from light; in solution store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.