Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylaluminum Bis(2,6-di-tert-butyl-4-methylphenoxide) (0.4mol/L in Toluene), TCI America™
SDP

Catalog No. M121150ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M121150ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M121150ML Supplier TCI America Supplier No. M121150ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56252-55-2
Molecular Formula C31H49AlO2
MDL Number MFCD00079652
Synonym Bis(2,6-di-tert-butyl-4-methylphenoxy)methylaluminum

Specifications

Quantity 50 mL
UN Number 3399
Formula Weight 480.71
Physical Form Liquid
Chemical Name or Material Methylaluminum Bis(2,6-di-tert-butyl-4-methylphenoxide) (0.4mol/L in Toluene)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.