Learn More
Medchemexpress LLC Methylamino-PEG1-Boc | 1807521-03-4 | 95.0% | 203.28 | 100 MG
SDP
Catalog No. 5000349484
Encompass_Preferred
Methylamino-PEG1-Boc, also known as Methylamino-PEG1-t-butyl ester, is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands joined by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. This mechanism allows PROTACs to harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • PROTACs consist of two distinct ligands joined by a linker
  • PROTACs harness the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Intended for research use only
Catalog No. 50-003-49484 Supplier Medchemexpress LLC Supplier No. HY140154100MG
Methylamino-PEG1-Boc, also known as Methylamino-PEG1-t-butyl ester, is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands joined by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. This mechanism allows PROTACs to harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • PROTACs consist of two distinct ligands joined by a linker
  • PROTACs harness the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Intended for research use only