Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylamino-PEG1-Boc | 1807521-03-4 | 95.0% | 203.28 | 250 MG
SDP

Catalog No. 5000352288
Encompass_Preferred

Methylamino-PEG1-Boc is a PEG-based PROTAC linker primarily used in the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs are designed with two different ligands connected by a linker: one targets an E3 ubiquitin ligase and the other targets a specific protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is intended for research use only.

  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight of 203.28
  • Formula C10H21NO3
  • Appears as a colorless to light yellow liquid

Catalog No. 50-003-52288 Supplier Medchemexpress LLC Supplier No. 250MGHY140154
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylamino-PEG1-Boc is a PEG-based PROTAC linker primarily used in the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs are designed with two different ligands connected by a linker: one targets an E3 ubiquitin ligase and the other targets a specific protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is intended for research use only.

  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight of 203.28
  • Formula C10H21NO3
  • Appears as a colorless to light yellow liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.