Learn More
Medchemexpress LLC Methylamino-PEG2-Boc | 1807521-04-5 | 100.0% | 247.33 | 1 G
SDP
Catalog No. 5000350521
Encompass_Preferred
Methylamino-PEG2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only and not sold to patients. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker.
  • Can be used in the synthesis of PROTACs.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
  • For research use only.
  • Appearance: colorless to light yellow oil.
  • Stable under recommended storage conditions.
Catalog No. 50-003-50521 Supplier Medchemexpress LLC Supplier No. HY1401551G
Methylamino-PEG2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only and not sold to patients. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker.
  • Can be used in the synthesis of PROTACs.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
  • For research use only.
  • Appearance: colorless to light yellow oil.
  • Stable under recommended storage conditions.