Learn More
Medchemexpress LLC Methylamino-PEG4-Boc | 1621616-14-5 | 335.44 | 1 G
SDP
Catalog No. 5000350309
Encompass_Preferred
Methylamino-PEG4-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • PROTACs selectively degrade target proteins
Catalog No. 50-003-50309 Supplier Medchemexpress LLC Supplier No. HY1401561G
Methylamino-PEG4-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • PROTACs selectively degrade target proteins