Learn More
Medchemexpress LLC Methylamino-PEG4-Boc | 1621616-14-5 | MFCD28155210 | ≥95% | 335.44 | 25 MG
SDP
Catalog No. 5000349975
Encompass_Preferred
Methylamino-PEG4-Boc is a polyethylene glycol (PEG)-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-49975 Supplier Medchemexpress LLC Supplier No. 25MGHY140156
Methylamino-PEG4-Boc is a polyethylene glycol (PEG)-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins