Learn More
Medchemexpress LLC Methylamino-PEG4-Boc | 1621616-14-5 | Pure form | 335.44 | 50 MG
SDP
Catalog No. 5000348006
Encompass_Preferred
Methylamino-PEG4-Boc is a PEG-based PROTAC linker primarily utilized in the synthesis of Proteolysis Targeting Chimeras (PROTACs). It acts as a versatile building block, enabling the construction of PROTAC molecules by connecting a ligand for an E3 ubiquitin ligase to a ligand for a target protein.
  • Used in the synthesis of PROTACs
  • PEG-based linker
  • Acts as a versatile building block for PROTAC molecules
Catalog No. 50-003-48006 Supplier Medchemexpress LLC Supplier No. HY14015650MG
Methylamino-PEG4-Boc is a PEG-based PROTAC linker primarily utilized in the synthesis of Proteolysis Targeting Chimeras (PROTACs). It acts as a versatile building block, enabling the construction of PROTAC molecules by connecting a ligand for an E3 ubiquitin ligase to a ligand for a target protein.
  • Used in the synthesis of PROTACs
  • PEG-based linker
  • Acts as a versatile building block for PROTAC molecules