Learn More
Medchemexpress LLC Methylarbutin | 6032-32-2 | 98.0% | 286.28 | 10 G
SDP
Catalog No. 5000452632
Encompass_Preferred
Methylarbutin is a chemical compound known for its anti-inflammatory and urinary antiseptic properties. It is commonly used in research settings to study these biological activities.
  • Exhibits anti-inflammatory activity
  • Possesses urinary antiseptic characteristics
  • Suitable for research applications
Catalog No. 50-004-52632 Supplier Medchemexpress LLC Supplier No. HYW00930410G
Add to Cart
Add to Cart
Methylarbutin is a chemical compound known for its anti-inflammatory and urinary antiseptic properties. It is commonly used in research settings to study these biological activities.
  • Exhibits anti-inflammatory activity
  • Possesses urinary antiseptic characteristics
  • Suitable for research applications