Learn More
Zymo Research Corporation Methylated-DNA IP Kit, 10 Rxns.
SDP
Catalog No. 501251558
Encompass_Preferred
The Methylated-DNA IP Kit enriches 5-methylcytosine (5-mC) containing DNA from any pool of fragmented genomic DNA for use in genome-wide methylation analysis.The kit includes a highly specific anti-5-methylcytosine monoclonal antibody for the “capture” and separation of methylated DNA from non-methylated DNA in only a fewhours. Typically, over a hundred-fold enrichment of methylated DNA vs. non-methylated DNA can be achieved with the use of this kit. Recovered DNA is suitable for many downstream applications to analyze genome-wide DNA methylation including: PCR, qPCR, whole-genome amplification, MeDIP-Seq, etc.
Catalog No. 50-125-1558 Supplier Zymo Research Corporation Supplier No. D5101
The Methylated-DNA IP Kit enriches 5-methylcytosine (5-mC) containing DNA from any pool of fragmented genomic DNA for use in genome-wide methylation analysis.The kit includes a highly specific anti-5-methylcytosine monoclonal antibody for the “capture” and separation of methylated DNA from non-methylated DNA in only a fewhours. Typically, over a hundred-fold enrichment of methylated DNA vs. non-methylated DNA can be achieved with the use of this kit. Recovered DNA is suitable for many downstream applications to analyze genome-wide DNA methylation including: PCR, qPCR, whole-genome amplification, MeDIP-Seq, etc.